The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008871.1 | NP_001008871.1 | 2 - 3 | 184 | 93 |
XM_006238095.2 | XP_006238157.1 | 3 - 4 | 1082 | 93 |