Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chmp5
Entrez Gene ID
297995
Gene Name
charged multivesicular body protein 5
Gene Aliases
RGD1305968
Chromosome Location
Chr.5: 57267312 - 57284455 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025410.1 NP_001020581.1 5 - 6 425 110
GenBank mRNA BC086333.1 - 4 - 5 287 110
BC097963.1 - 5 - 6 425 110
FQ214132.1 - 5 - 6 453 110
FQ234681.1 - 5 - 6 459 110

View Full Product Details