Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chmp5
Entrez Gene ID
297995
Gene Name
charged multivesicular body protein 5
Gene Aliases
RGD1305968
Chromosome Location
Chr.5: 57267312 - 57284455 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025410.1 NP_001020581.1 7 - 8 655 61
GenBank mRNA BC086333.1 - 6 - 7 517 61
BC097963.1 - 7 - 8 655 61
FQ214132.1 - 7 - 8 683 61
FQ228147.1 - 7 - 8 688 60
FQ234681.1 - 7 - 8 689 61

View Full Product Details