Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp6v0d2
Entrez Gene ID
297932
Gene Name
ATPase, H+ transporting, lysosomal V0 subunit D2
Gene Aliases
-
Chromosome Location
Chr.5: 33843591 - 33892446 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011972.1 NP_001011972.1 3 - 4 577 122
GenBank mRNA BC089917.1 - 3 - 4 577 122

View Full Product Details