Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snapc3
Entrez Gene ID
362537
Gene Name
small nuclear RNA activating complex, polypeptide 3
Gene Aliases
-
Chromosome Location
Chr.5: 101526629 - 101553335 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013212.1 NP_001013230.1 6 - 7 814 130
GenBank mRNA BC091186.1 - 6 - 7 814 130

View Full Product Details