Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdnf
Entrez Gene ID
361276
Gene Name
cerebral dopamine neurotrophic factor
Gene Aliases
Armetl1
Chromosome Location
Chr.17: 78721230 - 78735324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037543.1 NP_001032632.1 2 - 3 307 63

View Full Product Details