Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bend7
Entrez Gene ID
361275
Gene Name
BEN domain containing 7
Gene Aliases
RGD1305898
Chromosome Location
Chr.17: 77448430 - 77527888 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191834.1 NP_001178763.1 4 - 5 785 70

View Full Product Details