Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slain2
Entrez Gene ID
305310
Gene Name
SLAIN motif family, member 2
Gene Aliases
RGD1311593
Chromosome Location
Chr.14: 37802347 - 37871123 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107214.1 NP_001100684.1 8 - 9 1999 73
XM_006250898.2 XP_006250960.1 8 - 9 2074 73
XM_006250899.2 XP_006250961.1 7 - 8 1992 73
GenBank mRNA BC158815.1 - 6 - 7 1072 73

View Full Product Details