Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab32
Entrez Gene ID
365042
Gene Name
RAB32, member RAS oncogene family
Gene Aliases
RGD1559997
Chromosome Location
Chr.1: 4638663 - 4653220 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108902.1 NP_001102372.1 2 - 3 666 64

View Full Product Details