The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047856.2 | NP_001041321.2 | 3 - 4 | 646 | 115 |
GenBank mRNA | AY383685.1 | - | - | 567 | 115 |
FM118556.1 | - | 3 - 4 | 646 | 115 | |
FQ214391.1 | - | 3 - 4 | 632 | 115 | |
FQ220041.1 | - | 1 - 2 | 79 | 115 |