Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dlat
Entrez Gene ID
81654
Gene Name
dihydrolipoamide S-acetyltransferase
Gene Aliases
-
Chromosome Location
Chr.8: 55062549 - 55087832 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031025.1 NP_112287.1 8 - 9 1165 94
GenBank mRNA BC107440.1 - 8 - 9 1165 94
D00092.1 - 5 - 6 669 94
D10655.1 - 7 - 8 882 94
M16075.1 - 5 - 6 669 94

View Full Product Details