Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmg20a
Entrez Gene ID
315689
Gene Name
high mobility group 20A
Gene Aliases
RGD1564760
Chromosome Location
Chr.8: 61080103 - 61157613 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108150.1 NP_001101620.1 8 - 9 1181 67
XM_006243117.2 XP_006243179.1 8 - 9 4738 67

View Full Product Details