Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nlrx1
Entrez Gene ID
315599
Gene Name
NLR family member X1
Gene Aliases
RGD1311293
Chromosome Location
Chr.8: 48583559 - 48600203 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025010.1 NP_001020181.1 2 - 3 159 58
GenBank mRNA BC089834.1 - 2 - 3 159 58

View Full Product Details