Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Etfa
Entrez Gene ID
300726
Gene Name
electron-transfer-flavoprotein, alpha polypeptide
Gene Aliases
ETF
Chromosome Location
Chr.8: 60028786 - 60086352 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009668.1 NP_001009668.1 4 - 5 428 72
GenBank mRNA BC088412.1 - 4 - 5 428 72

View Full Product Details