Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyyr1
Entrez Gene ID
304138
Gene Name
cysteine/tyrosine-rich 1
Gene Aliases
-
Chromosome Location
Chr.11: 24967936 - 25078740 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013980.1 NP_001014002.1 2 - 3 466 64
GenBank mRNA BC087052.1 - 2 - 3 466 64

View Full Product Details