Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cers4
Entrez Gene ID
304208
Gene Name
ceramide synthase 4
Gene Aliases
Lass4
Chromosome Location
Chr.12: 4439750 - 4474729 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107117.1 NP_001100587.1 1 - 2 48 89

View Full Product Details