Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbx1
Entrez Gene ID
300084
Gene Name
ring-box 1, E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.7: 122700849 - 122710907 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034135.1 NP_001029307.1 - 320 152
GenBank mRNA BC100258.1 - - 320 152

View Full Product Details