Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nol12
Entrez Gene ID
362955
Gene Name
nucleolar protein 12
Gene Aliases
Nop25, RGD1305327
Chromosome Location
Chr.7: 120161238 - 120166852 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012747.2 NP_001012765.1 3 - 4 258 82
XM_006242007.2 XP_006242069.1 3 - 4 534 82
GenBank mRNA AY917132.1 - 3 - 4 236 82
BC103643.1 - 3 - 4 258 82
FQ227943.1 - 3 - 4 306 82

View Full Product Details