The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012747.2 | NP_001012765.1 | 4 - 5 | 404 | 86 |
XM_006242007.2 | XP_006242069.1 | 4 - 5 | 680 | 86 | |
GenBank mRNA | AY917132.1 | - | 4 - 5 | 382 | 86 |
BC103643.1 | - | 4 - 5 | 404 | 86 |