Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fkbp5
Entrez Gene ID
361810
Gene Name
FK506 binding protein 5
Gene Aliases
-
Chromosome Location
Chr.20: 7976704 - 8097291 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012174.1 NP_001012174.1 9 - 10 1132 84
XM_006256222.2 XP_006256284.1 9 - 10 1881 84
XM_006256224.2 XP_006256286.1 10 - 11 1948 84
GenBank mRNA BC085868.1 - 9 - 10 1132 84

View Full Product Details