The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008846.1 | NP_001008846.1 | 1 - 2 | 95 | 67 |
XM_006255970.2 | XP_006256032.1 | 1 - 2 | 297 | 67 | |
GenBank mRNA | KC222915.1 | - | 1 - 2 | 95 | 67 |
KC222930.1 | - | 1 - 2 | 95 | 67 |