Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgd2
Entrez Gene ID
309653
Gene Name
FYVE, RhoGEF and PH domain containing 2
Gene Aliases
-
Chromosome Location
Chr.20: 6973355 - 6990023 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107617.2 NP_001101087.2 5 - 6 864 100
XM_006256205.2 XP_006256267.1 5 - 6 910 100
XM_008772729.1 XP_008770951.1 5 - 6 910 100

View Full Product Details