Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
L2hgdh
Entrez Gene ID
314196
Gene Name
L-2-hydroxyglutarate dehydrogenase
Gene Aliases
RGD1306250
Chromosome Location
Chr.6: 92016560 - 92057643 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108028.1 NP_001101498.1 8 - 9 1078 138

View Full Product Details