Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mfsd2b
Entrez Gene ID
500624
Gene Name
major facilitator superfamily domain containing 2B
Gene Aliases
RGD1561716
Chromosome Location
Chr.6: 28956155 - 28968245 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729615.3 XP_002729661.1 5 - 6 862 59
XM_006239869.2 XP_006239931.1 5 - 6 889 59
XM_006239870.2 XP_006239932.1 4 - 5 763 59

View Full Product Details