Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcrl3
Entrez Gene ID
680697
Gene Name
Fc receptor-like 3
Gene Aliases
-
Chromosome Location
Chr.2: 186776470 - 186788834 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761269.1 XP_008759491.1 - 1044 96

View Full Product Details