Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lancl3
Entrez Gene ID
302540
Gene Name
LanC lantibiotic synthetase component C-like 3 (bacterial)
Gene Aliases
RGD1563642
Chromosome Location
Chr.X: 14358224 - 14490340 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191728.1 NP_001178657.1 3 - 4 900 98

View Full Product Details