Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw12
Entrez Gene ID
301015
Gene Name
F-box and WD repeat domain containing 12
Gene Aliases
RGD1307859
Chromosome Location
Chr.8: 117871409 - 117890930 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_236641.6 XP_236641.6 6 - 7 781 93

View Full Product Details