Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nlrc3
Entrez Gene ID
287070
Gene Name
NLR family, CARD domain containing 3
Gene Aliases
Nlrc3-ps1, RGD1304902
Chromosome Location
Chr.10: 11810889 - 11844184 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245834.2 XP_006245896.2 6 - 7 2342 66

View Full Product Details