Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec2e
Entrez Gene ID
689853
Gene Name
C-type lectin domain family 2, member E
Gene Aliases
Clec2d10
Chromosome Location
Chr.4: 162849880 - 162866382 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177689.1 NP_001171160.1 2 - 3 170 72
XM_008763336.1 XP_008761558.1 3 - 4 2262 72
GenBank mRNA FJ404473.1 - 2 - 3 206 72

View Full Product Details