Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam81b
Entrez Gene ID
309925
Gene Name
family with sequence similarity 81, member B
Gene Aliases
RGD1561830
Chromosome Location
Chr.2: 3004639 - 3124537 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760605.1 XP_008758827.1 7 - 8 880 145

View Full Product Details