Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mien1
Entrez Gene ID
360617
Gene Name
migration and invasion enhancer 1
Gene Aliases
RGD1306682
Chromosome Location
Chr.10: 86391848 - 86393141 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108296.1 NP_001101766.1 2 - 3 228 139
GenBank mRNA FQ214212.1 - - 225 139

Gene Symbol
LOC102552549
Entrez Gene ID
102552549
Gene Name
migration and invasion enhancer 1-like
Gene Aliases
-
Chromosome Location
Chr.10: 84846256 - 84847855 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247244.2 XP_006247306.1 2 - 3 541 139

View Full Product Details