The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012029.1 | NP_001012029.1 | 6 - 6 | 986 | 88 |
GenBank mRNA | BC085891.1 | - | 6 - 6 | 986 | 88 |