Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cep68
Entrez Gene ID
289822
Gene Name
centrosomal protein 68
Gene Aliases
RGD1309101
Chromosome Location
Chr.14: 104499907 - 104523326 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_214106.7 XP_214106.4 7 - 7 3340 146

View Full Product Details