Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cops7b
Entrez Gene ID
363273
Gene Name
COP9 signalosome subunit 7B
Gene Aliases
-
Chromosome Location
Chr.9: 93607171 - 93633352 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108807.1 NP_001102277.1 3 - 3 462 139
XM_006245414.2 XP_006245476.1 2 - 2 913 139
XM_006245415.2 XP_006245477.1 2 - 2 550 139

View Full Product Details