Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Plin1
- Entrez Gene ID
-
25629
- Gene Name
- perilipin 1
- Gene Aliases
-
PERIA, Plin
- Chromosome Location
-
Chr.1: 141458993 - 141471096 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
L26044.1 |
- |
7 - 7 |
1577 |
108 |
View Full Product Details