Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcs1l
Entrez Gene ID
301514
Gene Name
BC1 (ubiquinol-cytochrome c reductase) synthesis-like
Gene Aliases
-
Chromosome Location
Chr.9: 81868265 - 81872198 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007666.1 NP_001007667.1 3 - 4 771 122
GenBank mRNA BC083660.1 - 3 - 4 771 122

View Full Product Details