Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl16
Entrez Gene ID
494223
Gene Name
F-box and leucine-rich repeat protein 16
Gene Aliases
Scirr1
Chromosome Location
Chr.10: 15176471 - 15188726 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009504.1 NP_001009504.1 3 - 4 1227 122
XM_006246020.2 XP_006246082.1 4 - 5 1545 122
GenBank mRNA AY823669.1 - 3 - 4 1227 122

View Full Product Details