Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom1r78
Entrez Gene ID
494273
Gene Name
vomeronasal 1 receptor 78
Gene Aliases
V1rc36
Chromosome Location
Chr.4: 87943152 - 87944069 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008924.1 NP_001008924.1 1 - 1 325 165
GenBank mRNA AY510292.1 - 1 - 1 325 165

View Full Product Details