The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009531.1 | NP_001009531.1 | 1 - 1 | 630 | 151 |
GenBank mRNA | AY510330.1 | - | 1 - 1 | 630 | 151 |