Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klk1c6
Entrez Gene ID
408242
Gene Name
kallikrein 1-related peptidase C6
Gene Aliases
Gk11, Gk6, Klk6, rGK-6
Chromosome Location
Chr.1: 99947714 - 99964824 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003977.1 NP_001003977.1 1 - 2 53 142

View Full Product Details