The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007635.2 | NP_001007636.2 | 26 - 27 | 3947 | 129 |
XM_006232134.2 | XP_006232196.1 | 28 - 29 | 4289 | 129 | |
GenBank mRNA | AY631396.1 | - | 26 - 27 | 3947 | 129 |
AY631398.1 | - | 18 - 19 | 2886 | 129 | |
AY631399.1 | - | 26 - 27 | 3712 | 129 | |
AY631400.1 | - | 22 - 23 | 3086 | 129 |