The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024759.1 | NP_001019930.1 | 5 - 6 | 1091 | 134 |
XM_006241397.2 | XP_006241459.1 | 5 - 6 | 1156 | 134 | |
XM_008765391.1 | XP_008763613.1 | 4 - 5 | 843 | 134 | |
GenBank mRNA | BC097355.1 | - | 5 - 6 | 1091 | 134 |