The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025758.1 | NP_001020929.1 | 3 - 4 | 436 | 58 |
XM_008768093.1 | XP_008766315.1 | 3 - 4 | 342 | 58 | |
GenBank mRNA | BC098036.1 | - | 3 - 4 | 436 | 58 |