The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037548.1 | NP_001032637.1 | 2 - 2 | 150 | 74 |
GenBank mRNA | AY621339.1 | - | 2 - 2 | 150 | 74 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006253371.2 | XP_006253433.1 | 2 - 2 | 191 | 74 |