Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cntnap5b
Entrez Gene ID
301650
Gene Name
contactin associated protein-like 5B
Gene Aliases
Caspr5-2, Caspr5-3, Cntnap5c, RGD1565194
Chromosome Location
Chr.13: 21678088 - 22591058 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047873.1 NP_001041338.1 4 - 5 526 80
XM_008769451.1 XP_008767673.1 4 - 5 3275 80
XM_008769452.1 XP_008767674.1 4 - 5 959 80

View Full Product Details