Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd34
Entrez Gene ID
305081
Gene Name
CD34 molecule
Gene Aliases
-
Chromosome Location
Chr.13: 113691841 - 113711259 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107202.2 NP_001100672.1 2 - 3 256 114
XM_008769857.1 XP_008768079.1 2 - 3 546 114
GenBank mRNA EU448292.1 - 2 - 3 256 114
EU448293.1 - 2 - 3 256 114

View Full Product Details