Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf43
Entrez Gene ID
303412
Gene Name
ring finger protein 43
Gene Aliases
RGD1305204
Chromosome Location
Chr.10: 74956889 - 75028951 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135921.1 NP_001129393.1 2 - 3 380 89
XM_006247090.2 XP_006247152.1 3 - 4 1865 89
XM_006247091.2 XP_006247153.1 3 - 4 1640 89
XM_006247092.2 XP_006247154.1 2 - 3 2385 89
XM_008768038.1 XP_008766260.1 2 - 3 2776 89

View Full Product Details