Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf43
Entrez Gene ID
303412
Gene Name
ring finger protein 43
Gene Aliases
RGD1305204
Chromosome Location
Chr.10: 74956889 - 75028951 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135921.1 NP_001129393.1 7 - 8 952 108
XM_006247090.2 XP_006247152.1 8 - 9 2437 108
XM_006247091.2 XP_006247153.1 8 - 9 2212 108
XM_006247092.2 XP_006247154.1 7 - 8 2957 108
XM_006247094.2 XP_006247156.1 6 - 7 2313 108
XM_008768038.1 XP_008766260.1 7 - 8 3348 108
XM_008768039.1 XP_008766261.1 5 - 6 2991 108
XM_008768040.1 XP_008766262.1 6 - 7 2893 108
XM_008768041.1 XP_008766263.1 6 - 7 2541 108

View Full Product Details