Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf43
Entrez Gene ID
303412
Gene Name
ring finger protein 43
Gene Aliases
RGD1305204
Chromosome Location
Chr.10: 74956889 - 75028951 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135921.1 NP_001129393.1 8 - 9 2309 91

View Full Product Details