Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Zfp958
Entrez Gene ID
100302405
Gene Name
zinc finger protein 958
Gene Aliases
Zinki
Chromosome Location
Chr.12: 5485691 - 5492254 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142758.1 NP_001136230.1 - 1687 109
GenBank mRNA DQ490055.1 - - 1687 109

View Full Product Details